missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JT010
GREENER_CHOICE

Product Code. 18795616
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18795616

Brand: Tocris Bioscience™ 6269/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective TRPA1 channel activator (EC50 = 65 nM)

JT010 is a potent and selective TRPA1 channel activator (EC50 = 0.65 nM); activates the TRPA1 channel by covalently and site-selectively binding to Cys621. Exhibits selectivity for TRPA1 over TRPV1, TRPV3, TRPV4, TRPM2, TRPM8, and TRPC5 channels (EC50 values are >1 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-N-[4-(4-methoxyphenyl)-2-thiazolyl]-N-(3-methoxypropyl)acetamide
CAS 917562-33-5
Quantity 25 mg
Molecular Formula C16H19ClN2O3S
Formula Weight 354.85
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.