missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JTC 801

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
High affinity, selective NOP receptor antagonist (Ki = 8.2 nM). Displays approximately 12.5-, 129- and 1055-fold selectivity over human μ-, κ- and δ-opioid receptors respectively. Exhibits anti-nociceptive effects in acute pain models in vivo. Orally active.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 447.96 |
| Purity | >98% |
| Molecular Weight (g/mol) | 447.96 |
| Chemical Name or Material | JTC 801 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction