missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JTV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10424-1-AP-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against full-length of JTV1.
The JTV1 gene is located on chromosome 7p22 flanked by two genes, HRI and PMS2. JTV1 and HRI overlap slightly and are arranged in a tail-to-tail fashion. JTV1 and PMS2 are separated by approximately 200 base pairs and are arranged head-to-head. JTV1 is transcribed in the opposite direction compared to HRI and PMS2. The function of the JTV1 gene product is unknown.Specifications
| JTV1 | |
| Polyclonal | |
| Unconjugated | |
| AIMP2 | |
| AIMP2, JTV1, JTV1 gene, P38, PRO0992, Protein JTV 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 231872, 288480, 7965 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13155, Q32PX2, Q8R010 | |
| Aimp2 | |
| JTV1 Fusion Protein Ag0675 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction