missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jumonji/JARID2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2214MFV450
This item is not returnable.
View return policy
Description
Jumonji/JARID2 Polyclonal antibody specifically detects Jumonji/JARID2 in Human, Mouse, Rabbit samples. It is validated for Western Blot, Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP assay (ChIP), KnockDown
Specifications
| Jumonji/JARID2 | |
| Polyclonal | |
| Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation (ChIP), Knockout Validated | |
| JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji | |
| A synthetic peptide made to an N-terminal portion of the human JARID2 protein sequence (between residues 1-100). [UniProt# Q92833] | |
| 0.1 mL | |
| Chromatin Research, Epigenetics, Neuroscience, Transcription Factors and Regulators | |
| 3720 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Simple Western, ChIP Assay, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), ChIP Assay, Gene Knock-Out | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction