missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunB/AP-1 Antibody (PCRP-JUNB-3G11), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08390C
This item is not returnable.
View return policy
Description
JunB/AP-1 Monoclonal antibody specifically detects JunB/AP-1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| JunB/AP-1 | |
| Monoclonal | |
| DyLight 650 | |
| activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B | |
| Recombinant full-length human JunB/AP-1 protein (Uniprot: P17275) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence, Protein Array | |
| PCRP-JUNB-3G11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 3726 | |
| Store at 4°C in the dark. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction