missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JunB/AP-1 Mouse anti-Human, Clone: PCRP-JUNB-3G2, Novus Biologicals™
Mouse Monoclonal Antibody
€ 508.00
Specifications
| Antigen | JunB/AP-1 |
|---|---|
| Clone | PCRP-JUNB-3G2 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
JunB/AP-1 Monoclonal specifically detects JunB/AP-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| JunB/AP-1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 3726 | |
| IgG1 | |
| Protein A or G purified |
| PCRP-JUNB-3G2 | |
| Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| activator protein 1, AP-1, jun B proto-oncogene, transcription factor jun-B | |
| Recombinant full-length human JunB/AP-1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title