Learn More
3M™ Jupiter™ Powered Air Turbo Filters
Use with the Jupiter PAPR. The 3M Jupiter™ Powered Air Turbo Filters are available in a wide selection of gas or particulate protection.
Brand:  3M™ 7000145610
Additional Details : Weight : 0.77400kg
 View more versions of this product
Product Code. 10367649
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Filters are to be used with the Jupiter Powered Air Turbo respirator.
Specifications
Specifications
ABEP3
Combined Gas & Vapor and Particulate
Jupiter™ Powered-Air Respirator
12/Pk.