missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Pop-Up Drum Fill Gauge

Prevents overfillin when dispensing waste into drums

€ 29.50 - € 30.20


Products 2
Product Code Brand Length (English) Length (Metric) Price Quantity & Availability  
Product Code Brand Length (English) Length (Metric) Price Quantity & Availability  
15330103
View Documents Promotion Details
Justrite™
08530
5 in. 12.7 cm
€ 30.20
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
 
11964957
View Documents Promotion Details
Justrite™
08531
11 in. 27.94 cm
€ 29.50
Each
View alternative products
Save Up to 
»
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
 
Description

Description

  • Features handy indicator that pops up when drum is almost full
  • Nonmetallic
  • Fits 0.75 in. (1.9cm) bung
  • Available in two lengths

Videos
SDS
Documents

Documents

Product Certifications