missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ JWH 133

Product Code. 18014862
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18014862 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18014862 Supplier Tocris Bioscience™ Supplier No. 1343/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective CB2 agonist

JWH 133 is a potent CB2 selective agonist (Ki = 3.4 nM). Approx. 200-fold selective over CB1 receptors. Active in vivo, reducing spasticity in a murine model of multiple sclerosis. Activity also enhances the release of IL-10 by LPS/IFN-γ-stimulated macrophages and results in downregulation of the IL-12 subunit p40.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (6aR,10aR)-3-(1,1-Dimethylbutyl)-6a,7,10,10a-tetrahydro-6,6,9-trimethyl-6H-dibenzo[b,d]pyran
CAS 259869-55-1
Quantity 10 mg
Target Cannabinoid R2/CB2 Agonists
Molecular Formula C22H32O
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.