Learn More
Tocris Bioscience™ JX 06
GREENER_CHOICE
Potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor
Brand:  Tocris Bioscience™ 5845/10
Encompass
Product Code. 15797136
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
JX 06 is a potent and selective pyruvate dehydrogenase kinase (PDK) 1/2/3 inhibitor (IC50 values are 49, 101 and 313 nM for PDK1, PDK2 and PDK3, respectively). Exhibits selectivity for PDK1/2/3 over PDK4 and a panel of 323 other kinases. Suppresses A549 lung cancer cell proliferation in vitro. Reduces tumor volume of A549 xenografts in mice.
Specifications
Specifications
Bis(morpholinothiocarbonyl) disulfide
729-46-4