missing translation for 'onlineSavingsMsg'
Learn More

K-7174 dihydrochloride, MedChemExpress

Product Code. 16475308
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16475308

Brand: Medchem Express HY12743A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress K-7174 dihydrochloride is a novel cell adhesion inhibitor; inhibits the expression of vascular cell adhesion molecule-1 (VCAM-1) induced by either IL-1β or TNF-α.

K-7174 dihydrochloride is a novel cell adhesion inhibitor

Specifications

Chemical Name or Material K-7174 dihydrochloride
CAS 191089-60-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C33H50Cl2N2O6
Quantity 10 mg
Solubility Information H2O : 15 mg/mL (23.38 mM; Need ultrasonic and warming)
SMILES COC1=C(OC)C(OC)=CC(/C=C/CCCN2CCN(CCC/C=C/C3=CC(OC)=C(OC)C(OC)=C3)CCC2)=C1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 641.67
Formula Weight 641.67
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.