missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ K-Ras (G12A Mutant) Recombinant Rabbit Monoclonal Antibody (HL1153)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546940
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene, a Kirsten ras oncogene homolog from the mammalian ras gene family, encodes a protein that is a member of the small GTPase superfamily A single amino acid substitution is responsible for an activating mutation The transforming protein that results is implicated in various malignancies, including lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas and colorectal carcinoma Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region.
Specifications
| K-Ras (G12A Mutant) | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| ALPS4; C-BAS/HAS; CFC2; C-HA-RAS1; C-H-RAS; c-Ki-ras; c-Ki-ras2; c-K-ras; CMNS; CTLO; HAMSV; HRas proto-oncogene, GTPase; HRAS1; H-RASIDX; Ki-Ras; KRAS; K-Ras; KRAS proto-oncogene, GTPase; KRAS1; KRAS2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; NCMS; N-ras; NRAS proto-oncogene, GTPase; NRAS1; NS; NS3; NS6; p21ras; RALD; RASH1; RASK2 | |
| Carrier-protein conjugated synthetic peptide surrounding mutant G12A of human K-Ras. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1153 | |
| Unconjugated | |
| P29466, P48729, Q8N752 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 122011, 1452, 834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction