missing translation for 'onlineSavingsMsg'
Learn More
Learn More
K15 (a putative transmembrane protein similar to LMP2A) Antibody (IF3/A10) - BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
K15 (a putative transmembrane protein similar to LMP2A) Monoclonal antibody specifically detects K15 (a putative transmembrane protein similar to LMP2A) in Human, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
Specifications
| Antigen | K15 (a putative transmembrane protein similar to LMP2A) |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | IF3/A10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Recombinant protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?