Learn More
Robin™ Kahn's Rackless Shaker
Product Code. 11999607
Change view
Click to view available options
Speed Range:
275 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range unitSize
11999607 275 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11999607 Supplier Robin™ Supplier No. PA131
Kahn's Rackless Shakers are ideal for mixing, blending or agitating substances in a tube in a back-and-forth motion. Used with three racks and the optional plexiglass cover, the shaker will accept hemolysis tubes or BK tubes.
  • Back-and-forth shaker
  • Very quiet
  • Holds 90 hemolysis tubes (3 racks 49045) or 16 BK tubes (2 racks 49046)
  • Optional, dust-proof Plexiglas cover (sold separately)
  • 1 to 60 min. timer with continuous ON position
  • Compulsory accessories: 3 racks, and plexiglass cover
Specifications
Load Bearing Capacity (Metric) 6.5 kg
Speed Range 275 rpm
Electrical Requirements 230 V, 50 Hz, 25 W
Voltage 230 V
Wattage 25 W
Product Type Kahn Shaker
Frequency 50 Hz
Depth (Metric) 260 mm
Height (Metric) 190 mm
Length (Metric) 360 mm
Timer Range 1 to 60 min.