Learn More
Invitrogen™ Kaiso Recombinant Rabbit Monoclonal Antibody (24GB3670)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557324
 View more versions of this product
Product Code. 30275183
  • € 146.00 / 20µL
Description
Kaiso Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a transcriptional regulator with bimodal DNA-binding specificity, which binds to methylated CGCG and also to the non-methylated consensusKAISO-binding site TCCTGCNA. The protein contains an N-terminal POZ/BTB domain and 3 C-terminal zinc finger motifs. It recruits the N-CoR repressor complex to promote histone deacetylation and the formation of repressive chromatin structures in target gene promoters. It may contribute to the repression of target genes of the Wnt signaling pathway, and may also activate transcription of a subset of target genes by the recruitment of catenin delta-2 (CTNND2). Its interaction with catenin delta-1 (CTNND1) inhibits binding to both methylated and non-methylated DNA. It also interacts directly with the nuclear import receptor Importin-alpha 2 (also known as karyopherin alpha2 or RAG cohort 1), which may mediate nuclear import of this protein. Alternatively spliced transcript variants encoding the same protein have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kaiso
Recombinant Monoclonal
Unconjugated
ZBTB33
AW260253; E130014G12Rik; Kaiso; kaiso transcription factor; RGD1566309; RP23-322E15.9; Transcriptional regulator Kaiso; WUGSC:H_DJ525N14.1; Zbtb33; zinc finger and BTB domain containing 33; zinc finger and BTB domain-containing protein 33; zinc finger transcription factor Kaiso; ZNF348; ZNF-kaiso
Rabbit
Affinity chromatography
RUO
10009, 501506
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3670
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q86T24
ZBTB33
A synthesized peptide derived from human KAISO (aa 620-672).
20 μL
Primary
Human, Rat