missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAL1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76580UV
This item is not returnable.
View return policy
Description
KAL1 Polyclonal antibody specifically detects KAL1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| KAL1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| Adhesion molecule-like X-linked, ADMLXHHA, anosmin-1, KAL, KALIG1, KALIG-1, Kallmann syndrome 1 sequence, Kallmann syndrome interval gene 1, Kallmann syndrome protein, Kallmann syndrome-1 sequence (anosmin-1), KMS | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human Anosmin. The immunogen is located within amino acids 430 - 480 of Anosmin. Amino Acid Squence: KVYWKKTEDPTVNRYHVR | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Neuroscience | |
| 3730 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction