missing translation for 'onlineSavingsMsg'
Learn More

Kallikrein 1 Rabbit anti-Human, Clone: 105, Novus Biologicals™

Product Code. 18209047 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18209047 50 μg 50µL
18223647 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18209047 Supplier Novus Biologicals Supplier No. NBP28947450UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Kallikrein 1 Monoclonal specifically detects Kallikrein 1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 1
Applications ELISA
Classification Monoclonal
Clone 105
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias EC 3.4.21, EC 3.4.21.35, glandular kallikrein 1, hK1, kallikrein 1, kallikrein 1, renal/pancreas/salivary, kallikrein serine protease 1, kallikrein-1, Kidney/pancreas/salivary gland kallikrein, Klk6, KLKR, Tissue kallikrein
Host Species Rabbit
Immunogen Recombinant Human Kallikrein 1 Protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3816
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.