Learn More
Invitrogen™ Kallikrein 10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109887
Description
Immunogen sequence: LDPEAYGSPC ARGSQPWQVS LFNGLSFHCA GVLVDQSWVL TAAHCGNKPL WARVGDDHLL LLQGEQLRRT TRSVVHPKYH QGSGPILPRR TDEHDLMLLK LARPVVLGPR VRALQLPYRC AQPGDQCQVA GWGTTAARRV KYNKGLTCSS ITILSPKECE VFYPGVVTNN MICAGLDRGQ DPCQSDSGGP LVCDETLQGI LSWGVYPCGS AQHPAVYTQI CKYMSWINKV IRSN.
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its encoded protein is secreted and may play a role in suppression of tumorigenesis in breast and prostate cancers. Alternate splicing of this gene results in multiple transcript variants encoding the same protein.
Specifications
| Kallikrein 10 | |
| Polyclonal | |
| Unconjugated | |
| KLK10 | |
| 2300002A13Rik; breast normal epithelial cell associated serine protease; kallikrein 10; kallikrein 10 protein 1; kallikrein 10 protein 12; kallikrein 10 protein 13; kallikrein 10 protein 2; kallikrein 10 protein 3; kallikrein 10 protein 4; kallikrein 10 protein 5; kallikrein 10 protein 7; kallikrein 10 protein 8; kallikrein 10 protein 9; kallikrein related peptidase 10; kallikrein related-peptidase 10; kallikrein-10; kallikrein-related peptidase 10; Klk10; NES1; Normal epithelial cell-specific 1; protease serine-like 1; protease, serine-like, 1; PRSSL1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 292850, 5655, 69540 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 6.88 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43240, P36375 | |
| KLK10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 43-276 of human KLK10 (NP_6658951). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.