missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 12 Monoclonal Antibody (364932), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA524110
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Kallikrein 12 Monoclonal antibody specifically detects Kallikrein 12 in Human samples. It is validated for Immunoprecipitation, Neutralization, Western Blot.
Specifications
| Kallikrein 12 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9UKR0 | |
| KLK12 | |
| Mouse myeloma cell line NS0-derived recombinant human Kallikrein 12 Ala18-Asn248 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Neutralization, Western Blot | |
| 364932 | |
| Unconjugated | |
| KLK12 | |
| 2310008B01Rik; AI324852; kallikrein 12; kallikrein related peptidase 12; kallikrein related-peptidase 12; kallikrein-12; Kallikrein-like protein 5; kallikrein-related peptidase 12; KLK12; KLK-12; KLKL5; KLK-L5; UNQ669/PRO1303 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 43849 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction