missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 13 Antibody (001), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89329AF647
This item is not returnable.
View return policy
Description
Kallikrein 13 Monoclonal antibody specifically detects Kallikrein 13 in Human samples. It is validated for ELISA
Specifications
| Kallikrein 13 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 001 | |
| ELISA | |
| DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Kallikrein 13 (NP_056411.1; Met1-Ile262). | |
| 0.1 mL | |
| Cancer | |
| 26085 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction