missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 13 Antibody (MM0428-5X29), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11748F
This item is not returnable.
View return policy
Description
Kallikrein 13 Monoclonal antibody specifically detects Kallikrein 13 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Kallikrein 13 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MM0428-5X29 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp586J1923, EC 3.4.21, EC 3.4.21.-, kallikrein 13, Kallikrein-like protein 4, kallikrein-related peptidase 13, KLK-L4kallikrein-13, KLKL4kallikrein-like gene 4 | |
| Human recombinant Kallikrein 13 | |
| 0.1 mL | |
| Cancer | |
| 26085 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction