missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 15 Antibody (05), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26080FR
This item is not returnable.
View return policy
Description
Kallikrein 15 Monoclonal antibody specifically detects Kallikrein 15 in Human samples. It is validated for ELISA
Specifications
| Kallikrein 15 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 05 | |
| ELISA | |
| ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, prostinogen | |
| Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Kallikrein 15 (Accession#: NP_059979.2, Met1-Asn256) | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 55554 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction