Learn More
Kallikrein 15 Antibody (05), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26080IR
Product Code. 30031874
  • € 518.00 / 0.10mL
Description
Kallikrein 15 Monoclonal antibody specifically detects Kallikrein 15 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kallikrein 15
Monoclonal
DyLight 755
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
ELISA
05
ELISA
ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, prostinogen
Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Kallikrein 15 (Accession#: NP_059979.2, Met1-Asn256)
0.1 mL
Cancer, Tumor Suppressors
55554
Store at 4C in the dark.