missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 15 Antibody (MM0429-5Z12), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11749AF750
This item is not returnable.
View return policy
Description
Kallikrein 15 Monoclonal specifically detects Kallikrein 15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Kallikrein 15 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| KLK15 | |
| Human recombinant Kallikrein 15 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0429-5Z12 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, prostinogen | |
| Mouse | |
| Protein G purified | |
| Cancer, Tumor Suppressors | |
| 55554.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido