missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 2 Antibody (JNJ-69086420) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28696JF635
This item is not returnable.
View return policy
Description
Kallikrein 2 Monoclonal antibody specifically detects Kallikrein 2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Kallikrein 2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2 | |
| KLK2/ Kallikrein 2 | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 3817 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| JNJ-69086420 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction