Learn More
Novus Biologicals™ Kallikrein 2 Antibody (JNJ-69086420), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28696-100ug
 View more versions of this product
Product Code. 30047955
  • ONLINE EXCLUSIVE PRICE
    € 361.00
    € 341.25 / 100µg
    Save € 19.75
    5% Off
Description
Kallikrein 2 Monoclonal antibody specifically detects Kallikrein 2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kallikrein 2
Monoclonal
Unconjugated
P20151
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
JNJ-69086420
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
EC 3.4.21, EC 3.4.21.35, glandular kallikrein 2, Glandular kallikrein-1, hGK-1, hK2, kallikrein 2, prostatic, kallikrein-2, kallikrein-related peptidase 2, KLK2, KLK2A2, MGC12201, Tissue kallikrein-2
KLK2/ Kallikrein 2
100 μg
Cancer, Signal Transduction
3817