missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Kallikrein 4/Prostase/EMSP1 Antibody (KLK4/8942) [HRP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27013H
This item is not returnable.
View return policy
Description
Kallikrein 4/Prostase/EMSP1 Monoclonal antibody specifically detects Kallikrein 4/Prostase/EMSP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Kallikrein 4/Prostase/EMSP1 | |
| Monoclonal | |
| HRP | |
| AI2A1, ARM1, EC 3.4.21, EC 3.4.21.-, EMSP, EMSP1MGC116827, Enamel matrix serine proteinase 1, kallikrein 4 (prostase, enamel matrix, prostate), kallikrein-4, Kallikrein-like protein 1, kallikrein-related peptidase 4, KLK-L1MGC116828, Prostase, PRSS17enamel matrix serine protease 1, PSTSandrogen-regulated message 1, Serine protease 17 | |
| Recombinant fragment of human Kallikrein 4/Prostase/EMSP1 protein (around aa 1-200) (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 9622 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| KLK4/8942 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction