missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kallikrein 6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16306844
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16306844 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16306844 Supplier Invitrogen™ Supplier No. PA578134

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Target recombinant protein Predicted Reactivity: Dog (88%), Rhesus Monkey (98%), Bovine (83%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Kallikreins, including Kallikrein 6, are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. The KLK6 enzyme is regulated by steroid hormones. In tissue culture, the enzyme has been found to generate amyloidogenic fragments from the amyloid precursor protein, suggesting a potential for involvement in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kallikrein 6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Klk6
Gene Accession No. Q92876
Gene Alias 0610007D04Rik; AI849898; brain serine protease; BSP; Bssp; Glandular kallikrein K1; hK6; Kal; KAL-B; kallikrein 1; kallikrein 29; kallikrein 6; kallikrein 6 (neurosin); kallikrein 6 (neurosin, zyme); kallikrein 7; kallikrein related peptidase 6; kallikrein related-peptidase 6; kallikrein renal/pancreas/salivary; kallikrein-1; Kallikrein6; Kallikrein-6; Klk1; Klk1b6; Klk29; Klk6; Klk-6; Klk7; MGC9355; mGK-6; MSP; myelencephalon-specific protease; myofibril-bound protease; neurosin; Protease M; protease serine 9 (neurosin); protease, serine, 18; protease, serine, 9; PRSS18; PRSS9; renal kallikrein; Serine protease 18; serine protease 9; SP59; tissue kallikrein-6; true tissue kallikrein; zyme
Gene Symbols Klk6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Kallikrein 6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5653
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.