missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 8/Neuropsin Antibody (MM0434-6G15), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-11754F
This item is not returnable.
View return policy
Description
Kallikrein 8/Neuropsin Monoclonal antibody specifically detects Kallikrein 8/Neuropsin in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Kallikrein 8/Neuropsin | |
| Monoclonal | |
| FITC | |
| EC 3.4.21, EC 3.4.21.118, hK8, HNP, kallikrein 8 (neuropsin/ovasin), kallikrein-related peptidase 8, KLK8, KLK8 protein type 1, KLK8 protein type 2, Neuropsin, neuropsin type 1, neuropsin type 2, NP, NRPN, Ovasin, PRSS19kallikrein-8, Serine protease 19, Serine protease TADG-14, TADG14neuropsin, Tumor-associated differentially expressed gene 14 protein | |
| Human recombinant Kallikrein 8/Neuropsin | |
| 0.1 mL | |
| Neuroscience | |
| 11202 | |
| Store at 4°C in the dark. | |
| IgG2 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0434-6G15 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction