Learn More
Kanamycin disulfate, 700μg/mg, Thermo Scientific Chemicals
Product Code. 15428389
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428389 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15428389 Supplier Thermo Scientific Alfa Aesar Supplier No. J66856.06
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 64013-70-3 | C18H40N4O19S2 | 680.647 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64013-70-3
Molecular Formula C18H40N4O19S2
Molecular Weight (g/mol) 680.647
MDL Number MFCD00143916
InChI Key OGTKIXVMLDAMNU-KNQICTBBSA-N
Synonym Kanamycin A; Kanamycin acid sulfate
PubChem CID 24721551
IUPAC Name (2R,3S,4S,5R,6R)-2-(aminomethyl)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O.OS(=O)(=O)O
Specifications
Type Kanamycin Disulfate
Physical Form Powder
Melting Point >260°C
Color Colorless
Quantity 5 g
Sensitivity Light and moisture sensitive
Solubility Information Soluble in aqueous solutions
Formula Weight 680.65
Chemical Name or Material Kanamycin disulfate, 700μg/mg