missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KANK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553357
This item is not returnable.
View return policy
Description
Immunogen sequence: SQHIPTAEGA SGSNTEEEIR MELSPDLISA CLALEKYLDN PNALTERELK VAYTTVLQEW LRLACRSDAH PELVRRHLVT FRAMSARLLD YVVNIADSNG NTALHYSVSH ANFPVVQ Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
This gene encodes a member of the KN motif and ankyrin repeat domains (KANK) family of proteins, which play a role in cytoskeletal formation by regulating actin polymerization. The encoded protein functions in the sequestration of steroid receptor coactivators and possibly other proteins. Mutations in this gene are associated with impaired kidney podocyte function and nephrotic syndrome, and keratoderma and woolly hair.
Specifications
| KANK2 | |
| Polyclonal | |
| Unconjugated | |
| KANK2 | |
| AI504612; ANKRD25; ankyrin repeat domain 25; ankyrin repeat domain-containing protein 25; BC010245; Kank2; KIAA1518; kidney ankyrin repeat-containing protein 2; KN motif and ankyrin repeat domain-containing protein 2; KN motif and ankyrin repeat domains 2; matrix-remodeling-associated protein 3; MXRA3; PPKWH; RGD1563532; SIP; SRC-1-interacting protein; SRC1-interacting protein; SRC-interacting protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 25959 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q63ZY3 | |
| KANK2 | |
| Recombinant protein corresponding to Human KANK2. Recombinant protein control fragment (Product #RP-90786). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction