missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KANSL2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97893JF646
This item is not returnable.
View return policy
Description
KANSL2 Polyclonal specifically detects KANSL2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| KANSL2 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| C12orf41, chromosome 12 open reading frame 41, FLJ12670, FLJ20436, hypothetical protein LOC54934, KANSL2, KAT8 regulatory NSL complex subunit 2, NSL2 | |
| Produced in rabbits immunized with E. coli-derived Human KANSL2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 54934 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction