missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KAO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583176
This item is not returnable.
View return policy
Description
Immunogen sequence: DLSNQELKAV HSFLWSKKEL RLQPSSTTTM AKNTVFLIEM LLPKKYHVLR FLDKGERHPV REARAVIFFG DQEHPNVTEF A Highest antigen sequence indentity to the following orthologs: Mouse - 73%, Rat - 73%.
This gene encodes a metal-binding membrane glycoprotein that oxidatively deaminates putrescine, histamine, and related compounds. The encoded protein is inhibited by amiloride, a diuretic that acts by closing epithelial sodium ion channels. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| KAO | |
| Polyclonal | |
| Unconjugated | |
| AOC1 | |
| 1600012D06Rik; Abp; abp1; amiloride binding protein 1 (amine oxidase (copper-containing)); amiloride binding protein 1 (amine oxidase, copper-containing); amiloride binding protein/diamine oxidase; amiloride-binding protein 1; amiloride-sensitive amine oxidase; amiloride-sensitive amine oxidase [copper-containing]; amine oxidase copper containing 1; Amine oxidase copper domain-containing protein 1; amine oxidase, copper containing 1; amine oxidase, copper-containing 1; aoc1; DAO; DAO1; Diamine oxidase; histaminase; KAO; kidney amine oxidase; si:ch211-286c5.2; zgc:154101 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P19801 | |
| AOC1 | |
| Recombinant protein corresponding to Human AOC1. Recombinant protein control fragment (Product #RP-94963). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction