missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAP1, p Ser824 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32073G
This item is not returnable.
View return policy
Description
KAP1 Polyclonal antibody specifically detects KAP1 in Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)
Specifications
| KAP1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation (Negative) | |
| E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28 | |
| The immunogen this antibody was made to, maps to a region of mouse KAP-1 (tripartite motif-containing 28 ) surrounding serine 824 using the numbering given in entry NP_035718.2 (Gene ID 21849) when the serine residue is phosphorylated. | |
| 0.1 mL | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
| 10155 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction