Learn More
KAP1 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-15202-100UL
Description
The KAP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human KAP1. This antibody recognizes human antigen. The KAP1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specifications
| KAP1 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human |
| Flow Cytometry (Intracellular Staining) | |
| CoraLite 488 | |
| 100 μL | |
| 10155 | |
| IgG |