Learn More
Kappa Light Chain Antibody (HP6053), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34639DL594
Product Code. 18625189
  • ONLINE EXCLUSIVE PRICE
    € 563.00
    € 532.35 / 0.10mL
    Save € 30.65
    5% Off
Description
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Kappa Light Chain
Monoclonal
DyLight 594
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809
Purified human Kappa Light Chain
0.1 mL
Adaptive Immunity, Immunology
3514
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
HP6053
50mM Sodium Borate
Mouse
Protein A or G purified