missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Antibody (KLC709), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34660AF647
This item is not returnable.
View return policy
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Product Image.
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| Alexa Fluor 647 | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Mouse | |
| 22.5 kDa | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| KLC709 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| IGKC | |
| Recombinant human Ig kappa chain | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| This monoclonal antibody is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. It recognizes human Ig kappa light chains of both secreted and cell surface immunoglobulin. It detects also free kappa light chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant. | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction