missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Kappa Light Chain Recombinant Rabbit Monoclonal Antibody (JE56-54)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544906
This item is not returnable.
View return policy
Description
Kappa Light Chain Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Immunoglobulin classes share the same basic four polypeptide chain structure of two heavy chains (five types) and two light chains (kappa, lambda; both having a molecular weight of 22.5 kDa). Kappa and lambda consist of a variable region and a constant region and can easily be differentiated by the antigenic properties of the constant region. The ratio of kappa to lambda is 70:30. Antibodies to the kappa light chain are reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Specifications
| Kappa Light Chain | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P01601, P01602, P01834, P0DOX7 | |
| IGK, IGKC, IGKV1-5, IGKV1D-16 | |
| Full length native human IgG. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE56-54 | |
| Unconjugated | |
| IGK | |
| D4Mit8; HCAK1; ig kappa chain c region; Igk; Igk@; Igkc; Igk-C; IGKCD; IGKV; IGKV15; IGKV1-5; Immunoglobulin K; immunoglobulin kappa chain complex; immunoglobulin kappa constant; immunoglobulin kappa light chain; immunoglobulin kappa locus; immunoglobulin kappa variable 1-5; kappa; Kappa LC; Km; L12; L12a; LOC500183; similar to NGF-binding Ig light chain; V1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 28299, 28901, 3514, 50802 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction