missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ KAPSTO™ GPN 735 Sealing Screw Plugs
Click to view available options
Length (Metric):
18 mm
Unit Size:
Pack of 100
Description
- Installation with either screw driver or spanner
- Particularly suitable for processes like cleaning, testing, painting or transport
- For closing internal threads acc. with DIN EN 3852-1 and DIN ISO 6149-1
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | For closing internal threads in accordance with DIN 3852-1, DIN ISO 6149-1 |
| Length (Metric) | 18 mm |
| Quantity | 100 Pack |
| Material | Polyamide |
| Type | Sealing Plug |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction