Learn More
Poeppelmann™ Kapsto™ Polyethylene Flange Cover
Product Code. 10053794
Click to view available options
Diameter (Metric):
102 mm
280 mm
Unit Size:
Pack of 100
Pack of 25
Product Code. 10053794
Brand: Poeppelmann™ 67001500000
Only null left
Add to basket
Add to basket
Protects against coarse impurities. Poppelmann Kapsto™ Polyethylene Flange Cover is perfect for accommodating major tolerances.
  • Polyethylene
  • Yellow
Specifications
Diameter (Metric) 280 mm
Material Polyethylene
Overall Length (Metric) 34 mm
For Use With Protect against coarse impurities, prevent damage to the sealing face and close off screw holes
For Use With (Application) Protect against coarse impurities, prevent damage to the sealing face and close off screw holes
Packaging Quantity 25 Pack
Color Yellow
Product Type Flange Cover