missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ Kapsto™ Polyethylene Flange Cover

Product Code. p-8000381
Click to view available options
Diameter (Metric):
102 mm
280 mm
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 10155254

Brand: Poeppelmann™ 65100500000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Protects against coarse impurities. Poppelmann Kapsto™ Polyethylene Flange Cover is perfect for accommodating major tolerances.

  • Polyethylene
  • Yellow

Specifications

Diameter (Metric) 102 mm
Material Polyethylene
For Use With Perfect for accommodating major tolerances
For Use With (Application) Close the opening and protect the raised sealing face of flanged valves and fittings
Packaging Quantity 100 Pack
Color Yellow
Product Type Flange Cover
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.