Learn More
Poeppelmann™ Kapsto™ Polyethylene Flathead Closing Plugs, GPN300 Series
Product Code. 10472355
Change view
Click to view available options
Length (Metric):
9.5 mm
10.5 mm
18 mm
Size (Metric):
25 mm
28 mm
45 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Size (Metric) unitSize
10472355 10.5 mm 25 mm Pack of 100
10597194 9.5 mm 28 mm Pack of 100
10329204 18 mm 45 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10472355 Supplier Poeppelmann™ Supplier No. 30000190000
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
Manufactured from polyethylene. Poppelmann™ Kapsto™ Polyethylene Flathead Closing Plugs, GPN300 Series protect internal threads and close off bores.
  • Polyethylene
  • Natural color
Specifications
Color Natural
For Use With (Application) For protection of internal threads and for closing off bores
Length (Metric) 10.5 mm
Quantity 100 Pack
Size (Metric) 25 mm
Material Polyethylene
Type Closing Plug