Learn More
Poeppelmann™ Kapsto™ Polyethylene Flathead Closing Plugs, GPN300 Series
Product Code. 10597194
Change view
Click to view available options
Length (Metric):
9.5 mm
10.5 mm
18 mm
Size (Metric):
25 mm
28 mm
45 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Size (Metric) Unit Size
10597194 9.5 mm 28 mm Pack of 100
10472355 10.5 mm 25 mm Pack of 100
10329204 18 mm 45 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10597194 Supplier Poeppelmann™ Supplier No. 30002320000
  • ONLINE EXCLUSIVE PRICE
    € 17.97
    € 11.32 / Pack of 100
    Save € 6.65
    37% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from polyethylene. Poppelmann™ Kapsto™ Polyethylene Flathead Closing Plugs, GPN300 Series protect internal threads and close off bores.
  • Polyethylene
  • Natural color
Specifications
Color Natural
For Use With (Application) For protection of internal threads and for closing off bores
Length (Metric) 9.5 mm
Quantity 100 Pack
Size (Metric) 28 mm
Material Polyethylene
Type Closing Plug