missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ Kapsto™ Polyethylene Protective Caps

Product Code. 10441795
Click to view available options
Overall Length (Metric):
20 mm
35 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10441795

Brand: Poeppelmann™ 25001600000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from polyethylene. Poppelmann™ Kapsto™ Polyethylene Protective Caps protect DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127 pipes during transport and storage.

  • Polyethylene
  • Yellow
  • Protect pipes during transport and storage

Specifications

Material Polyethylene
Overall Length (Metric) 20 mm
For Use With (Application) For protection of DIN EN 10220, DIN EN 10255 and DIN EN ISO 1127 pipes during transport and storage
Packaging Quantity 100 Pack
Color Yellow
Product Type Protective Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.