Learn More
Poeppelmann™ Kapsto™ Polyethylene Screw Plugs with Metric Threading, GPN700 Series
Click to view available options
Diameter (Metric) Outside:
16 mm
19 mm
21 mm
31.5 mm
Size (Metric):
G 1/4 Thread
G 3/4 Thread
G 3/8 Thread
M12x1 Thread
Unit Size:
Pack of 100
Description
- Polyethylene
- Yellow
Specifications
Specifications
| Color | Yellow |
| For Use With (Application) | Suitable for a wide range of metric and imperial threads |
| Quantity | 100 Pack |
| Size (Metric) | G 1/4 Thread |
| Diameter (Metric) Outside | 19 mm |
| Material | Polyethylene |
| Type | Screw Plugs |