Learn More
Poeppelmann™ Kapsto™ Polyethylene Screw Plugs with Metric Threading, GPN700 Series
Manufactured from polyethylene. Poppelmann™ Kapsto™ Polyethylene Screw Plugs with Metric Threading, GPN700 Series are suitable for a wide range of metric and imperial threads.
€ 28.40 - € 59.50
Specifications
Color | Yellow |
---|---|
Material | Polyethylene |
Type | Screw Plugs |
For Use With (Application) | Suitable for a wide range of metric and imperial threads |
Quantity | 100 Pack |
Product Code | Brand | Length (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Description | Price | Quantity & Availability | ||||
10063604
|
Poeppelmann™
70004000000 |
14 mm | With G 1/4 Threading |
€ 31.25
Pack of 100 |
In Stock
Log in to see stock. |
||||
10575484
|
Poeppelmann™
70033710000 |
20 mm | With G 3/4 Threading |
€ 59.50
Pack of 100 |
Estimated Shipment: 22-05-2024
Log in to see stock. |
||||
10239954
|
Poeppelmann™
70012100000 |
14 mm | With M12 Threading |
€ 28.40
Pack of 100 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
10309094
|
Poeppelmann™
70003860000 |
16 mm | With G 3/8 Threading |
€ 38.05
Pack of 100 |
Estimated Shipment: 24-05-2024
Log in to see stock. |
||||
Description
- Polyethylene
- Yellow
Specifications
Yellow | |
Screw Plugs | |
100 Pack |
Polyethylene | |
Suitable for a wide range of metric and imperial threads |