Learn More
Poeppelmann™ Kapsto™ Tapered Universal Protection Plugs, GPN600 Series
Click to view available options
Length (Metric):
11.2 mm
25 mm
30.6 mm
42.5 mm
Size (Metric):
15 mm
30 mm
35 mm
51.5 mm
Unit Size:
Pack of 100
Description
- Polyethylene
- Natural color
Specifications
Specifications
| Color | Natural |
| For Use With (Application) | Use as plugs in bores or internal thread, or as caps for external thread |
| Length (Metric) | 42.5 mm |
| Quantity | 100 Pack |
| Size (Metric) | 51.5 mm |
| Material | Polyethylene |
| Type | Cone Closures |