Learn More
Poeppelmann™ Kapsto™ Tapered Universal Protection Plugs, GPN600 Series
Product Code. 10788365
Change view
Click to view available options
Length (Metric):
11.2 mm
25 mm
30.6 mm
42.5 mm
Size (Metric):
15 mm
30 mm
35 mm
51.5 mm
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Size (Metric) unitSize
10788365 11.2 mm 15 mm Pack of 100
10348574 25 mm 30 mm Pack of 100
10033624 42.5 mm 51.5 mm Pack of 100
10788185 30.6 mm 35 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10788365 Supplier Poeppelmann™ Supplier No. 60001140000
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Manufactured from polyethylene. Poppelmann™ Kapsto™ Tapered Universal Protection Plugs, GPN600 Series can be used as plugs in bores or internal thread, or as caps for external thread.
  • Polyethylene
  • Natural color
Specifications
Color Natural
For Use With (Application) Use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 11.2 mm
Quantity 100 Pack
Size (Metric) 15 mm
Material Polyethylene
Type Cone Closures