Learn More
Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs
Product Code. 10738405
Change view
Click to view available options
Diameter (Metric) Outside:
17 mm
18 mm
19 mm
21 mm
22 mm
24 mm
29 mm
31.7 dia. x 22mmH
36.6 dia. x 23mmH
37 mm
39.7 dia. x 17mmH
39.7 dia. x 19mmH
42.8 dia. x 19mmH
7.5 dia. x 10mmH
8.3 dia. x 10.0mmH
Diameter (Metric) Inside:
11.3 mm
12.5 mm
13.5 mm
14 mm
15 mm
16.3 mm
17.3 mm
17.5 mm
22 mm
29.8 mm
31.4 dia. x 31mmH
33.3 dia. x 22mmH
38.2 dia. x 23mmH
41.4 dia. x 19mmH
44.4 dia. x 19mmH
9.2 dia. x 10mmH
9.9 dia. x 10.0mmH
Height (Metric):
10 mm
12 mm
13 mm
14 mm
15 mm
16 mm
38 mm
41 mm
48 mm
49 mm
51 mm
Unit Size:
Pack of 100
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Diameter (Metric) Inside Height (Metric) Unit Size
10738405 39.7 dia. x 19mmH 41.4 dia. x 19mmH 49 mm Pack of 100
10686635 17 mm 11.3 mm 10 mm Pack of 100
10328624 18 mm 12.5 mm 12 mm Pack of 100
10093514 19 mm 13.5 mm 12 mm Pack of 100
10618175 19 mm 14 mm 12 mm Pack of 100
10257024 21 mm 15 mm 13 mm Pack of 100
10628175 22 mm 17.3 mm 13 mm Pack of 100
10451405 24 mm 17.5 mm 15 mm Pack of 100
10587754 22 mm 16.3 mm 13 mm Pack of 100
10105504 29 mm 22 mm 16 mm Pack of 100
10537794 37 mm 29.8 mm 16 mm Pack of 100
10043384 39.7 dia. x 17mmH 31.4 dia. x 31mmH 38 mm Pack of 100
10155694 31.7 dia. x 22mmH 33.3 dia. x 22mmH 41 mm Pack of 100
10470255 36.6 dia. x 23mmH 38.2 dia. x 23mmH 48 mm Pack of 100
10388564 7.5 dia. x 10mmH 9.2 dia. x 10mmH 14 mm Pack of 100
10758575 42.8 dia. x 19mmH 44.4 dia. x 19mmH 51 mm Pack of 100
10155584 8.3 dia. x 10.0mmH 9.9 dia. x 10.0mmH 15 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 10738405 Supplier Poeppelmann™ Supplier No. 61000320000
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 610 Universal Protection Caps/Plugs are manufactured from yellow PE-LD plastic.
  • Universal use as a plug or cap
  • Acc. with VDA Band 19 and ISO 16232
  • Tapered design ideal for accomodating tolerances
Specifications
Color Yellow
Diameter (Metric) Inside 41.4 dia. x 19mmH
For Use With (Application) Used universally as plug or cap, Tapered design is ideal for accommHodating tolerances
Height (Metric) 49 mm
Length (Metric) 43 mm
Quantity 100 Pack
Diameter (Metric) Outside 39.7 dia. x 19mmH
Material Polyethylene
Type Closing Plug