missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KARS Recombinant Rabbit Monoclonal Antibody (JE40-55)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534815
This item is not returnable.
View return policy
Description
Positive Control: Cytoplasm.Cell membrane. Mitochondrion. Nucleus. Secreted.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| KARS | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q15046 | |
| KARS1 | |
| Recombinant protein within Human Lysyl tRNA synthetase aa 460-597. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| JE40-55 | |
| Unconjugated | |
| KARS1 | |
| AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3735 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction